missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyoD1 (Rhabdomyosarcoma Marker) Mouse, Clone: rMYD712, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 4654-MSM6-P1
Additional Details : Weight : 0.01000kg
Specifications
MyoD1 (Rhabdomyosarcoma Marker) | |
Nuclear. Only nuclear staining should be considered as evidence of skeletal muscle differentiation. | |
rMYD712 | |
Unconjugated | |
Mouse | |
45kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
4654 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human MyoD1 protein | |
Rhabdomyosarcoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |