missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyoD1 (Rhabdomyosarcoma Marker) Mouse, Clone: rMYD712, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | MyoD1 (Rhabdomyosarcoma Marker) |
---|---|
Clone | rMYD712 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15950548
|
EnquireBio
4654-MSM6-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940548
|
EnquireBio
4654-MSM6-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
MyoD1 (Rhabdomyosarcoma Marker) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human MyoD1 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
45kDa |
rMYD712 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
4654 | |
IgG1 κ | |
Protein A/G purified from Bioreactor concentrate. | |
Nuclear. Only nuclear staining should be considered as evidence of skeletal muscle differentiation. | |
Rhabdomyosarcoma |