Learn More
MYH15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557086
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KMERERADLT QDLADLNERL EEVGGSSLAQ LEITKKQETK FQKLHRDMEE ATLHFETTSA SLKKRH Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 73%.
MYH15 Polyclonal antibody specifically detects MYH15 in Human samples. It is validated for Immunohistochemistry.Specifications
MYH15 | |
Polyclonal | |
Unconjugated | |
MYH15 | |
KIAA1000; MYH15; Myosin heavy chain 15; myosin, heavy chain 15; myosin, heavy polypeptide 15; myosin-15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22989 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2K3 | |
MYH15 | |
Recombinant protein corresponding to Human MYH15 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only