missing translation for 'onlineSavingsMsg'
Learn More

Myeloperoxidase (MPO), PE, clone: MPO455-8E6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1299-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15516876

  • £75.29 / 25 tests


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The monoclonal antibody MPO455-8E6 recognizes myeloperoxidase (MPO), a protein within the azurophilic granules of myeloid cells. MPO is a multimeric protein comprised of two 55kDa subunits and two 15kDa subunits. The larger subunits associate with a heme protein resulting in a greenish color. As an enzyme, MPO breaks down hydrogen peroxide and oxidizes tyrosine. The products of this reaction, hypochlorous acid and tyrosyl radical, cause the cytotoxic and killing effects characteristic of neutrophils. Myeloperoxidase is important in the diagnosis of some cancers and increases in serum levels have been shown to correlate with cardiac events.Applications Reported: This MPO455-8E6 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This MPO455-8E6 antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells using the Intracellular Fixation & Permeablization Buffer Set (cat. 88-8824). For best results, whole blood should first be stained with antibodies to surface antigens then treated with 1X RBC lysis buffer (cat. 00-4333) to lyse erythrocytes. Then, for intracellular staining for MPO, cells should be fixed with IC Fixation Buffer (cat. 00-8222) washed two times with Permeabilization Buffer (cat. 00-8333) and then incubated with MPO455-8E6 for 30-60 minutes. After washing, cells may be analyzed on a flow cytometer. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Myeloperoxidase (MPO)
MPO455-8E6
PE
Conjugated
IgG1
RUO
Primary
Human
Flow Cytometry (Intracellular Staining)
5μL (0.5μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos