Thermo Scientific™ myECL™ Imager Accessories and Replacement Parts

Document and analyze images of blots and gels using a sensitive and easy-to-use system with an intuitive touchscreen interface and advanced integrated software.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: EW84857

UNSPSC: 45121517

Code: PN

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 13408329

Quantity Price
1 £ 1307.3 / Each
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Specification

Formulation One year extended warranty
Sufficient For Covers all parts (non-consumable) and labor; includes shipping and handling
Description Extended Warranty
Quantity 1 warranty
Item Description Extended Warranty
For Use With (Application) myECL Imager

Replacement parts for the myECL Imager and accessories to expand upon its capabilities.