Learn More
Thermo Scientific™ myECL™ Imager Accessories and Replacement Parts
Document and analyze images of blots and gels using a sensitive and easy-to-use system with an intuitive touchscreen interface and advanced integrated software.
Brand:  Thermo Scientific™ 62238
Code : PN
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 13355071
  • £788.57 / Each
Estimated Shipment
08-03-2018
Add to basket
Description
Description
Replacement parts for the myECL Imager and accessories to expand upon its capabilities.
Specifications
Specifications
Orange Filter
565-620nm bandpass filter; threaded, 50mm-diameter glass filter
Replacement for the pre-installed orange filter in the myECL Imager for detection of colorimetric protein stains (silver, coomassie) and wavelengths of common fluorescent nucleic acid gel stains (e.g., ethidium bromide) and protein stains
myECL Imager
1 filter
Orange Filter
Videos