Learn More
MYCBP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559476
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LVALYEEPEK PNSALDFLKH HLGAATPENP EIELLRLELA EMKEKYEAIV EE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
MYCBP Polyclonal antibody specifically detects MYCBP in Human samples. It is validated for Immunohistochemistry.Specifications
MYCBP | |
Polyclonal | |
Unconjugated | |
MYCBP | |
5730488M09Rik; Amy1; AMY-1; Associate of Myc 1; associate of myc-1; AW552132; AW742590; c-myc binding protein; C-Myc-binding protein; hypothetical protein LOC539291; LOW QUALITY PROTEIN: C-Myc-binding protein; MYC binding protein; MYCBP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26292 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99417 | |
MYCBP | |
Recombinant protein corresponding to Human MYCBP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only