Learn More
Tocris Bioscience™ MY-5445
Brand:  Tocris Bioscience™ 043250
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13487643
  • £475.00 / 50mg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
Specific inhibitor of cyclic GMP phosphodiesterase, selective for PDE5 (IC50 = 0.5μM).
Specifications
Specifications
1-(3-Chlorophenylamino)-4-phenylphthalazine
C20H14ClN3
50mg
78351-75-4
Observed MW: 331.8
Soluble to 50 mM in DMSO and to 5 mM in ethanol
Videos