missing translation for 'onlineSavingsMsg'
Learn More

Muscimol, Tocris Bioscience™

Potent GABAA agonist; also GABAA-ρ partial agonist

Brand:  Tocris (Bio-Techne) 028950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13437533

  • £505.72 / 50mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent GABAA receptor agonist and partial GABAA-ρ receptor agonist. Inhibits memory retention and attenuates airway constriction in vivo.
Specifications

Chemical Identifiers

2763-96-4
114.104
muscimol, pantherine, agarin, agarine, 3 2h-isoxazolone, 5-aminomethyl, 5-aminomethyl-3-isoxyzole, pantherin, 5-aminomethyl-3-isoxazolol, 3-hydroxy-5-aminomethylisoxazole, 5-aminomethyl-3-hydroxyisoxazole
CHEBI:7035
C1=C(ONC1=O)CN
C4H6N2O2
ZJQHPWUVQPJPQT-UHFFFAOYSA-N
4266
5-(aminomethyl)-1,2-oxazol-3-one

Specifications

Muscimol
2763-96-4
muscimol, pantherine, agarin, agarine, 3 2h-isoxazolone, 5-aminomethyl, 5-aminomethyl-3-isoxyzole, pantherin, 5-aminomethyl-3-isoxazolol, 3-hydroxy-5-aminomethylisoxazole, 5-aminomethyl-3-hydroxyisoxazole
C1=C(ONC1=O)CN
114.104
CHEBI:7035
>98%
50mg
C4H6N2O2
ZJQHPWUVQPJPQT-UHFFFAOYSA-N
5-(aminomethyl)-1,2-oxazol-3-one
4266
114.1
Videos