Learn More
Musashi-2 Monoclonal Antibody (C1), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 50-9677-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Msi2 (musashi homolog 2), also known as MSI2H, is a 328 amino acid protein that localizes to the cytoplasm and contains two RRM (RNA recognition motif) domains. Expressed ubiquitously at low levels, Msi2 functions as an RNA binding protein that, by regulating the expression of target mRNAs, is thought to play a role in the proliferation and maintenance of stem cells within the central nervous system. Msi2 is subject to posttranslational phosphorylation and is upregulated in response to brain injury, suggesting a role in healing and brain tissue regeneration. Chromosomal aberrations involving the Msi2 gene are associated with the progression of chronic myeloid leukemia. Multiple isoforms of Msi2 exist due to alternative splicing events.Tissue specificity: Ubiquitous; detected at low levels.
Musashi-2 Monoclonal antibody specifically detects Musashi-2 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
Musashi-2 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q920Q6, Q96DH6 | |
MSI2 | |
Affinity chromatography | |
RUO | |
124540, 76626 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
C1 | |
eFluor 660 | |
MSI2 | |
1700105C15Rik; AI451722; AI563628; AW489193; MSI2; Msi2h; musashi homolog 2; musashi RNA binding protein 2; musashi RNA-binding protein 2; Musashi2; musashi-2; RGD1560397; RNA-binding protein Musashi homolog 2 | |
Mouse | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG3 κ |
For Research Use Only.