Learn More
Thermo Scientific™ Multisip™ Split Septum Plugs
Thermo Scientific™ MULTISIP™ Split Septum Plugs feature a unique proprietary split septum design
£715.00
Specifications
Material Inert polymer
Pierceable Yes
Sterility Non-sterile
View More Specs
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10039789 View Documents Thermo Scientific™
AB-1174
7mm; 10 Trays of 96 plugs £715.00
Pack of 960 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Individual plugs made from an inert polymer
  • Pierceable with standard pipetting equipment
  • 96 plugs can be pierced simultaneously
  • 100% resealing even after 200 piercings
  • Less than 4N pierce pressure
  • Solvent resistant, including acetonitrile and DMSO (for long-term storage with DMSO, samples should be stored at 4°C or below)
  • Temperature range -170°C to +120°C; fully autoclavable
  • Apply 96 plugs individually with the Cap-It tool
Specifications
Specifications
Inert polymer
Yes
Non-sterile