Learn More
Thermo Scientific™ Multifuge X3R TX-1000 Cell Culture Centrifuge Package
Process a range of general purpose applications including cell culture, microplate, bioproduction, blood separation and microbiology.
Brand:  Thermo Scientific™ 50144014
Code : NEW
Additional Details : Weight : 72.00000kg
Product Code. 15233457
  • £12869.45 / 1 unit
In Stock 2
Includes: Heraeus Multifuge X3R, 230V (Mfr PN: 75004515); TX-1000 rotor, Slide coat (Mfr PN: 75003017); TX-1000 buckets (set of 4) (Mfr PN: 75003001); TX-1000 ClickSeal biocontainment lids (set of 4) (Mfr PN: 75007309); Adapter, 50mL Conical (Mfr PN: 75003674); Adapter, 24 x 15mL. BIOFlex HC (Mfr PN: 75007306)
Description
Description
Featuring high throughput and energy-efficient operation, this series can process up to 40% more conical tubes in swing-out rotor configuration and 100% more in fixed-angle configuration.
Specifications
Specifications
Heraeus Multifuge X3R, 230V (Mfr PN: 75004515); TX-1000 rotor, Slide coat (Mfr PN: 75003017); TX-1000 buckets (set of 4) (Mfr PN: 75003001); TX-1000 ClickSeal biocontainment lids (set of 4) (Mfr PN: 75007309); Adapter, 50mL Conical (Mfr PN: 75003674); Adapter, 24 x 15mL. BIOFlex HC (Mfr PN: 75007306)
Multifuge X3R TX-1000 Cell Culture Centrifuge Package
Videos
Documents
Warranty