missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Multidrop™ Combi Reagent Dispenser

Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo Scientific™ Multidrop™ Combi Reagent Dispenser.

Brand:  Thermo Scientific™ 5840300

Code : 50

Additional Details : Weight : 23.00000kg

 View more versions of this product

Product Code. 11560725

  • £14499.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

The Multidrop Combi Reagent Dispenser handles a wide selection of plates and volume range and provides fast dispensing and high-throughput operation.

  • Provides precise dispensing over a 0.5 to 2500μL range, ensuring reproducible assay data
  • Accommodates microplates from 6 to 1536 wells and plate heights of 5 to 50mm
  • Visual icon-based graphic display makes it easy to use and program, even without training
  • Minimal dead volume and back-flushing reduces reagent costs
  • Easy to use Thermo Scientific FILLit Software provides increased flexibility and functionality
  • Full robotic compatibility gives increased throughput
  • Uses eight-channel detachable and autoclavable dispensing cassettes that are standard across the Multidrop range

Multidrop Combi SMART:

  • Adds an advanced built-in tracing system to the Multidrop Combi
  • Provides improved realiability and cassette lifetime traceability, greatly enhancing user efficiency and reporting capability

Includes:

One standard tube dispensing cassette and two small tube dispensing cassettes (Multidrop Combi cat. nos. 24072670, 24073290, 24073295 and Multidrop Combi SMART cat.nos. 24072675, 24073292 and 24073297)

Recommended for:

  • Assay development
  • Primary and secondary screening
  • Compound storage
  • Genomics and proteomics research
  • PCR set-up
  • Sequencing set-up
  • Cell based assays
  • Bead based assays
  • ELISA assays
Specifications

Specifications

6- to 1536-well plates
Small tube dispensing cassette: 2μL, ±10% (typical); 10μL, ±5% (typical); >10μL, ±5% (typical) Stand tube dispensing cassette: 5μL, ±3%(typical); 20μL, ±2% (typical); 100μL, ±1% (typical); >100μL, ±1% (typical)
96-well plate: 10µL in 3 sec.; 20µL in 4 sec.; 100µL in 10 sec
384-well plate: 1µL in 5 sec.; 5µL in 5 sec.; 10µL in 6 sec.; 20µL in 9 sec.
1536-well plate: 1µL in 14 sec.; 5µL in 26 sec.
<1mL volume range 0.5-50μL; <7mL volume range >50μL
9.1kg
22cm
Multidrop Combi
100 to 240V
0.5 to 2500µL
Small tube dispensing cassette: 2µL, ±10% (typical); 10µL, ±5% (typical); >10µL, ±5% (typical)
Stand tube dispensing cassette: 5µL, ±3%(typical); 20µL, ±2% (typical); 100µL, ±1% (typical); >100µL, ±1% (typical)
Small tube dispensing cassette: 0.5µL increments 0.5-50µL
Stand tube dispensing cassette: 5µL increments 5-2500µL
Serial RS-232, USB
35.5cm
33cm
50/60Hz
100 to 240V 50/60Hz
Videos
Documentation Instrument & Equipment Services