missing translation for 'onlineSavingsMsg'
Learn More

Gas Arc™ Multi Stage Gas Regulator

Regulators to BS EN ISO 9001 and BS EN ISO 2503

POA - £359.34

Specifications

Type Gas Regulator
View More Specs

Products 2
Product Code Brand Gases Measured Price Quantity & Availability  
Product Code Brand Gases Measured Price Quantity & Availability  
12960790
Encompass
View Documents Promotion Details
Gas Arc™
3960001001
Acetylene
£359.34
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12930820
Encompass
View Documents Promotion Details
Gas Arc™
3960002176
Fuel Gas
POA
Each
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

  • Regulators to BS EN ISO 9001 and BS EN ISO 2503
  • Two gauge single and multi stage
  • Cylinder pressure up to 300bar
  • Robust brass construction to meet industry requirements
  • Tamper proof high flow safety relief valve
  • Safety gauges to BS EN ISO 562
  • Encapsulated valve design fitted with 316L sintered stainless steel filter
  • Regulator outlet connection: 3/8 inch BSP
  • Propane regulator is plugged
Specifications

Specifications

Gas Regulator
Videos
SDS
Documents

Documents

Product Certifications