Learn More
Multi-Element QC-21 Check Standard Solution, Specpure™, Thermo Scientific Chemicals
£302.00 - £739.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11364038
|
Thermo Scientific Alfa Aesar
041328.AE |
100 mL |
£302.00
100mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11490381
|
Thermo Scientific Alfa Aesar
041328.AP |
500 mL |
£739.00
500mL |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Multi-Element QC-21 Check Standard Solution is used as a certified reference material or quality control check standard for inductively coupled plasma optical emission spectroscopy (ICP-OES). It is also used in inductively coupled plasma mass spectrometry (ICP-MS), and flame or furnace atomic absorption spectroscopy.