missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Multi-Element Plasma Standard Solution 2, Specpure™

Brand:  Alfa Aesar™ 042882.AE

Code : NEW

Additional Details : Weight : 0.00010kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Quantity 100mL

Product Code. 15466525

  • £343.80 / 100mL


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Specifications

Multi-Element Plasma Standard Solution 2
Liquid
UN3264
Matrix: dilute HNO3
Specpure™
MFCD00212338
100mL
Videos
SDS