missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MULT1 (NKG2D Ligand) Armenian Hamster anti-Mouse, Rat, Clone: 5D10, eBioscience™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 14-5863-81
Code : Z2
Additional Details : Weight : 0.01000kg
Description
The 5D10 monoclonal antibody reacts with the murine MULT-1 (Murine ULBP-Like Transcript 1). MULT-1 is a type I transmembrane protein with two Ig-like domain and is one of the known murine NKG2-D ligands that include RAE1 molecules (alpha, beta, epsilon, gamma and delta) and H60. MULT-1 is a high affinity ligand for NKG2D.Expression of NKG2-D ligands is low or absent on normal adult tissues. However, stressed or transformed cells express NKG2-D ligand and can activate NK cell tumoricidal activity. Until now, the expression of NKG2-D ligands has been mainly studied with NKG2-D tetramers recognizing all NKG2-D ligands.Specifications
MULT1 (NKG2D Ligand) | |
Monoclonal | |
0.5mg/mL | |
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
Armenian Hamster | |
RUO | |
Mouse, Rat | |
Purified |
Flow Cytometry, Immunohistochemistry (Frozen) | |
5D10 | |
Unconjugated | |
NKG2DL, MULT-1 | |
50 μg | |
Primary | |
Store at 2-8°C. | |
IgG |
For Research Use Only.