Learn More
Cole-Parmer™ Mucksucker Drum and Tank Sampler
High-suction syringe action is ideal for viscous fluids
Brand:  Cole-Parmer™ 99200-20
Additional Details : Weight : 1.00000kg
Product Code. 12776047
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Drum and tank sampler.
  • 3.5 ft. long.
  • Efficiently draws samples that are close to the bottom of the drum or tank.
  • Comes with both a push/seal bottom and a coring bottom.
  • Coring bottom features a wide mouth for handling sludge samples.
Specifications
Specifications
Drum and Tank Sampler
3.5 ft.