Thermo Scientific™ Mucin 2 (MUC2) Ab-2, Mouse Monoclonal Antibody

Deliver accurate, reproducible results with the Thermo Scientific™ Mucin 2 (MUC2) Ab-2, Mouse Monoclonal Antibody.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: MS-1037-R1

1 ML MONOCLONAL ANTIBODY, ANTI-MUCIN 2 (MUC2)Ab-2, hostmouse, clone M53, isotype IgG2a/k,

UNSPSC: 12352203

Code: LD

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12638696

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Specification

Applications Immunohistochemistry (Paraffin)
Antigen Mucin 2 (MUC2) Ab-2
Host Species Mouse
Isotype IgG2a, κ
Quantity 1mL
Regulatory Status IVD
Conjugate Unlabeled
Cross Reactivity Human
Format Prediluted
Formulation Ready-to-use for IHC (UVLP)
Immunogen Recombinant protein encoding 5 tandem repeats of the human MUC2 apomucin
Research Discipline Cancer and Tumor Biology
Type Primary
Classification Monoclonal
Clone M53