Learn More
MTPG, Tocris Bioscience™
Group II/group III mGlu antagonist. More selective for group II than group III
Brand:  Tocris (Bio-Techne) 085550
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13488073
  • £625.89 / 50mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Group II/group III metabotropic glutamate receptor antagonist, showing selectivity for group II in electrophysiological studies.
Specifications
Chemical Identifiers
169209-66-9
233.231
(RS)-α-Methyl-4-tetrazolylphenylglycine
C10H11N5O2
PUQSGMIEFYCTSW-UHFFFAOYSA-N
5311456
Specifications
MTPG
>98%
(RS)-α-Methyl-4-tetrazolylphenylglycine
CC(C1=CC=C(C=C1)N2C=NN=N2)(C(=O)O)N
233.231
233.23
169209-66-9
C10H11N5O2
PUQSGMIEFYCTSW-UHFFFAOYSA-N
2-amino-2-[4-(tetrazol-1-yl)phenyl]propanoic acid
5311456