Learn More
Tocris Bioscience™ MTEP hydrochloride
Potent and selective mGlu5 antagonist
Brand:  Tocris Bioscience™ 292150
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13432054
  • £652.68 / 50mg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
Potent, selective and non-competitive mGlu5 antagonist (IC50 = 5nM in Ca2+-flux assay; Ki= 16nM). Displays anxiolytic activity in vivo and is orally active.
Specifications
Specifications
3-((2-Methyl-1,3-thiazol-4-yl)ethynyl)pyridine hydrochloride
>99%
Observed MW: 236.72
Soluble to 100mM in DMSO
1186195-60-7
C11H8N2S.HCl