Learn More
MSPG, Tocris Bioscience™
Group II/group III mGlu antagonist
Brand:  Tocris (Bio-Techne) 0854/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13458073
  • £139.05 / 10mg
Estimated Shipment
06-03-2019
Add to basket
Description
Description
Relatively non-selective antagonist of presynaptic mGlu receptors in neonatal rat spinal cord and adult rat cerebrocortical mGlu receptors.
Specifications
Chemical Identifiers
169209-64-7
245.249
mspg, rs-mspg, 2-amino-2-4-sulfophenyl propanoic acid, +/--alpha-methyl-4-sulfonophenyl glycine, rs-?-methyl-4-sulfonophenylglycine, inverted exclamation marka-, a-methyl-4-sulfonophenyl glycine
2-amino-2-(4-sulfophenyl)propanoic acid
C9H11NO5S
MVDSFPIEJILRME-UHFFFAOYSA-N
3964410
CC(C1=CC=C(C=C1)S(=O)(=O)O)(C(=O)O)N
Specifications
MSPG
C9H11NO5S
MVDSFPIEJILRME-UHFFFAOYSA-N
2-amino-2-(4-sulfophenyl)propanoic acid
3964410
>98%
169209-64-7
mspg, rs-mspg, 2-amino-2-4-sulfophenyl propanoic acid, +/--alpha-methyl-4-sulfonophenyl glycine, rs-?-methyl-4-sulfonophenylglycine, inverted exclamation marka-, a-methyl-4-sulfonophenyl glycine
CC(C1=CC=C(C=C1)S(=O)(=O)O)(C(=O)O)N
245.249
245.25