Learn More
MSOP, Tocris Bioscience™
Specific group III mGlu antagonist
Brand:  Tocris (Bio-Techne) 0803/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13458023
  • £165.00 / 10mg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
Selective group III metabotropic glutamate receptor antagonist. Displays an apparent KD?of 51 μM for the L-AP4-sensitive presynaptic mGluR on primary afferent terminals in spinal cord compared to > 700 μM for the (1S,3S)-ACPD-sensitive presynaptic mGlu in the same system. Has no activity on postsynaptic mGlu receptors or on ionotropic glutamate receptors on neonatal rat motoneurons. Also available as part of the Group III mGlu Receptor Tocriset™ .
Specifications
Chemical Identifiers
66515-29-5
199.099
msop, alpha-msop, alpha-methylserine-o-phosphate, rs-alpha-methylserine-o-phosphate, d0o8ak, rs-?-methylserine-o-phosphate, 2-amino-2-methyl-3-phosphonooxy propanoic acid
2-amino-2-methyl-3-phosphonooxypropanoic acid
C4H10NO6P
GSFCOAGADOGIGE-UHFFFAOYSA-N
3964633
CC(COP(=O)(O)O)(C(=O)O)N
Specifications
MSOP
C4H10NO6P
GSFCOAGADOGIGE-UHFFFAOYSA-N
2-amino-2-methyl-3-phosphonooxypropanoic acid
3964633
>98%
66515-29-5
msop, alpha-msop, alpha-methylserine-o-phosphate, rs-alpha-methylserine-o-phosphate, d0o8ak, rs-?-methylserine-o-phosphate, 2-amino-2-methyl-3-phosphonooxy propanoic acid
CC(COP(=O)(O)O)(C(=O)O)N
199.099
199.1