missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MSIA™ D.A.R.T.'S™ for the Versette™ and Novus i Platform

Improve hybrid MS bio-analysis with this exclusive workflow solution for proteins and peptides.

Brand:  Thermo Scientific™ 991R

Code : NEW

Additional Details : Weight : 0.45000kg

 View more versions of this product

Product Code. 15192579

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Improve hybrid MS bio-analysis with Thermo Scientific™ MSIA™ D.A.R.T.'S™ for the Versette™ and Novus i Platform , an exclusive Thermo Fisher Scientific workflow solution for proteins and peptides. Enabled through proprietary MSIA D.A.R.T.'S™ technologies, these workflows have demonstrated pg/mL level detection while providing simultaneous qualitative/quantitative data content from complex biological matrix, such as human plasma. Ideal for biologic and biomarker bio-analysis, MSIA provides innovative enablement in the areas of PKPD, DMPK, ADME, Biotransformation (DAR and payload determination), as well as routine biomarker analysis.

Next Generation Hybrid Mass Spectrometry Bio-Analytical Workflows:
  • Enables analysis of proteins of interest down to femtomole level
  • Streamlined workflow for protein qualitative & quantative bio-analysis
  • Employs a proprietary molecular trapping micro-column technology in an easy-to-use pipette tip housing format
  • Drives antibody/antigen binding kinetics and greatly improves assaying of low-abundance protein from a biological sample
Enabling Workflows and Technologies for Biologic and Biomarker Research:
  • Immunoaffinity column captures protein of interest with improved signal-to-noise ratio
  • Superior specificity and assaying characteristics compared to conventional approaches
  • Allows for hybrid assaying down to femtomole levels
  • Streptavidin and Protein A, G & A/G MSIA D.A.R.T.'S provide end user flexibility to analyze their specific targets of interest
  • Custom MSIA D.A.R.T.'S are available upon request
  • Optimized workflow solution Insulin (endogenous and therapeutic forms)

Requires:

300μL pipette such as Thermo Scientific™ Versette™ Liquid Handling Platform, Thermo Scientific™ Finnpipette™ Novus i Multichannel pipetters (listed in user manual)

Recommended for:

Hybrid mass spectrometry based assays

For research use only
Specifications

Specifications

MSIA D.A.R.T.'S, Reloadable Rack
2° to 8°C
MSIA D.A.R.T.S
300 μL
Reload Rack (tips not included)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers