Learn More
Phospho-Histone H3 (Ser10) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 491041
Code : NEW
Additional Details : Weight : 0.02000kg
Description
Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post tranlationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
Phospho-Histone H3 (Ser10) Polyclonal antibody specifically detects Phospho-Histone H3 (Ser10) in Human samples. It is validated for ChIP Assay, Dot Blot, ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
Phospho-Histone H3 (Ser10) | |
Polyclonal | |
Unconjugated | |
H3C1 | |
BUR5; CG31613; CG31613-PA; CG33803; CG33806; CG33809; CG33812; CG33815; CG33818; CG33821; CG33824; CG33827; CG33830; CG33833; CG33836; CG33839; CG33842; CG33845; CG33848; CG33851; CG33854; CG33857; CG33860; CG33863; CG33866; CG5825; CG5825-PA; CG5825-PC; CG5825-PD; CG8989; dH3.3A; Dmel\CG31613; Dmel\CG5825; Dmel_CG31613; Dmel_CG5825; fb58e10; h3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3B.1; H3 histone, family 3C; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; h3.2a; H3.3; H3.3 histone A; H3.3 histone B; H3.3a; H3.3B; H3.5; H3.5 histone; H3.A; H3.B; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-143; H3-291; H3-3A; H3-3B; H3-5; H3-53; H3-614; H3a; H3b; H3-B; H3C1; H3c10; H3c11; H3C12; H3C2; H3C3; H3C4; H3C6 | |
Rabbit | |
Unpurified | |
RUO | |
126961, 333932, 8350, 8356, 8357, 8358, 8968 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ChIP Assay, Dot Blot, ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Conc. Not Determined | |
whole serum with <0.1% sodium azide | |
P68431, Q71DI3 | |
H3C1, H3C10, H3C12, H3C14, H3C15, H3C2, H3C7, HIST1H3A | |
raised against histone H3 containing the phosphorylated serine 10 (H3S10p), using a KLH-conjugated synthetic peptide, using a KLH-conjugated synthetic peptide | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |