missing translation for 'onlineSavingsMsg'
Learn More

MS 245 oxalate, Tocris Bioscience™

High affinity 5-HT6 antagonist

Brand:  Tocris (Bio-Techne) 420810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13444354

  • £149.85 / 10mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

High affinity 5-HT6 antagonist (Ki = 2.1 nM). Potentiates the hypolocomotor actions of (-)-nicotine in mice.
Specifications

Chemical Identifiers

275363-58-1
448.49
ms 245 oxalate, 2-5-methoxy-1-phenylsulfonyl-1h-indol-3-yl-n,n-dimethylethanamine oxalate, 5-methoxy-n,n-dimethyl-1-phenylsulfonyl-1h-indole-3-ethanamine oxalate, 2-1-benzenesulfonyl-5-methoxyindol-3-yl-n,n-dimethylethanamine; oxalic acid
2-[1-(benzenesulfonyl)-5-methoxyindol-3-yl]-N,N-dimethylethanamine;oxalic acid
C21H24N2O7S
BLSWAEGCRSFTJG-UHFFFAOYSA-N
6918541
CN(C)CCC1=CN(C2=C1C=C(C=C2)OC)S(=O)(=O)C3=CC=CC=C3.C(=O)(C(=O)O)O

Specifications

MS 245 oxalate
275363-58-1
ms 245 oxalate, 2-5-methoxy-1-phenylsulfonyl-1h-indol-3-yl-n,n-dimethylethanamine oxalate, 5-methoxy-n,n-dimethyl-1-phenylsulfonyl-1h-indole-3-ethanamine oxalate, 2-1-benzenesulfonyl-5-methoxyindol-3-yl-n,n-dimethylethanamine; oxalic acid
CN(C)CCC1=CN(C2=C1C=C(C=C2)OC)S(=O)(=O)C3=CC=CC=C3.C(=O)(C(=O)O)O
448.49
448.49
10mg
C21H24N2O7S
BLSWAEGCRSFTJG-UHFFFAOYSA-N
2-[1-(benzenesulfonyl)-5-methoxyindol-3-yl]-N,N-dimethylethanamine;oxalic acid
6918541
>99%
Videos