Learn More
Tocris Bioscience™ MRS 2578
Selective P2Y6 antagonist
Brand:  Tocris Bioscience™ 214650
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13460654
  • £396.52 / 50mg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
Selective antagonist of P2Y6 nucleotide receptors; IC50 values are 37 and 98 nM at human and rat P2Y6 receptors respectively. Displays no activity at P2Y1, P2Y2, P2Y4 and P2Y11 receptors (IC50 > 10 μM). Inhibits agonist-induced cardiomyocyte contraction and UDP-induced phagocytosis.
Specifications
Specifications
MRS 2578
MRS 2578
472.66