Learn More
MRS 2219, Tocris Bioscience™
Potentiates P2X1-mediated responses
Brand:  Tocris (Bio-Techne) 120350
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13428863
  • £501.27 / 50mg
Estimated Shipment date
from Supplier 25-03-2020
Description
Description
A selective potentiator of ATP-evoked responses at rat P2X1 receptors (EC50 = 5.9 μM).
Specifications
Chemical Identifiers
14141-47-0
231.144
chembl74121, 3,9-dihydroxy-8-methyl-1h,5h-3??-1,3,2 dioxaphosphepino 5,6-c pyridin-3-one, tocris-1203, d03eeo, 1,5-dihydro-3-hydroxy-8-methyl 1,3,2 dioxaphosphepino 5,6-c pyridin-9-ol-3-oxide, 1,3,2 dioxaphosphepino 5,6-c pyridin-9-ol,1,5-dihydro-3-hydroxy-8-methyl-,3-oxide, 3,9-dihydroxy-8-methyl-1,5-dihydro 1,3,2 dioxaphosphepino 5,6-c pyridine 3-oxide, 3-hydroxy-8-methyl-3-oxo-1,5-dihydro-1,3,2 dioxaphosphepino 5,6-c pyridin-9-ol, 3-methyl-7-oxo-5,9-dihydro-6,8-dioxa-2-aza-7lambda*5*-phospha-benzocycloheptene-4,7-diol, 4-hydroxy-10-methyl-4-oxo-3,5-dioxa-9-aza-4$l^ 5-phosphabicyclo 5.4.0 undeca-1 11 ,7,9-trien-11-ol
3-hydroxy-8-methyl-3-oxo-1,5-dihydro-[1,3,2]dioxaphosphepino[5,6-c]pyridin-9-ol
C8H10NO5P
KCAJHODRYGNBSX-UHFFFAOYSA-N
3960826
CC1=NC=C2COP(=O)(OCC2=C1O)O
Specifications
50mg
14141-47-0
chembl74121, 3,9-dihydroxy-8-methyl-1h,5h-3??-1,3,2 dioxaphosphepino 5,6-c pyridin-3-one, tocris-1203, d03eeo, 1,5-dihydro-3-hydroxy-8-methyl 1,3,2 dioxaphosphepino 5,6-c pyridin-9-ol-3-oxide, 1,3,2 dioxaphosphepino 5,6-c pyridin-9-ol,1,5-dihydro-3-hydroxy-8-methyl-,3-oxide, 3,9-dihydroxy-8-methyl-1,5-dihydro 1,3,2 dioxaphosphepino 5,6-c pyridine 3-oxide, 3-hydroxy-8-methyl-3-oxo-1,5-dihydro-1,3,2 dioxaphosphepino 5,6-c pyridin-9-ol, 3-methyl-7-oxo-5,9-dihydro-6,8-dioxa-2-aza-7lambda*5*-phospha-benzocycloheptene-4,7-diol, 4-hydroxy-10-methyl-4-oxo-3,5-dioxa-9-aza-4$l^ 5-phosphabicyclo 5.4.0 undeca-1 11 ,7,9-trien-11-ol
CC1=NC=C2COP(=O)(OCC2=C1O)O
231.144
231.14
MRS 2219
C8H10NO5P
KCAJHODRYGNBSX-UHFFFAOYSA-N
3-hydroxy-8-methyl-3-oxo-1,5-dihydro-[1,3,2]dioxaphosphepino[5,6-c]pyridin-9-ol
3960826
>99%