Learn More
Tocris Bioscience™ MRS 2211
Brand:  Tocris Bioscience™ 240250
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13461144
  • £396.52 / 50mg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Competitive P2Y13 receptor antagonist (pIC50 = 5.97) that displays > 20-fold selectivity over P2Y1 and P2Y12 receptors.
Specifications
Specifications
2-[(2-Chloro-5-nitrophenyl)azo]-5-hydroxy-6-methyl-3-[(phosphonooxy)methyl]-4-pyridinecarboxaldehyde disodium salt
C14H10N4O8Na2PCl
50mg
>96%
Observed MW: 474.66
Soluble to 70mM in water