missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MRS 2211
Competitive P2Y13 antagonist
POA
Specifications
Chemical Name or Material | 2-[(2-Chloro-5-nitrophenyl)azo]-5-hydroxy-6-methyl-3-[(phosphonooxy)methyl]-4-pyridinecarboxaldehyde disodium salt |
---|---|
Assay Percent Range | >96% |
Solubility | Soluble to 70mM in water |
Molecular Formula | C14H10N4O8Na2PCl |
Formula Weight | Observed MW: 474.66 |
Description
Competitive P2Y13 receptor antagonist (pIC50 = 5.97) that displays > 20-fold selectivity over P2Y1 and P2Y12 receptors.Specifications
2-[(2-Chloro-5-nitrophenyl)azo]-5-hydroxy-6-methyl-3-[(phosphonooxy)methyl]-4-pyridinecarboxaldehyde disodium salt | |
Soluble to 70mM in water | |
Observed MW: 474.66 |
>96% | |
C14H10N4O8Na2PCl |
For Research Use Only