missing translation for 'onlineSavingsMsg'
Learn More

MRS 1754, Tocris Bioscience™

Selective A2B antagonist

Brand:  Tocris (Bio-Techne) 275250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13440524

  • £684.77 / 50mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective adenosine A2B receptor antagonist (Ki values are 1.97, 16.8, 403, 503, 570 and 612 nM for hA2B, rA1, hA1, hA2A, hA3 and rA2A receptors respectively).
Specifications

Chemical Identifiers

264622-58-4
486.532
n-4-cyanophenyl-2-4-2,6-dioxo-1,3-dipropyl-7h-purin-8-yl phenoxy acetamide, n-4-cyanophenyl-2-4-2,3,6,7-tetrahydro-2,6-dioxo-1,3-dipropyl-1h-purin-8-yl phenoxy-acetamide, lopac-m-6316, d01qyq, d0ne5p, atl-gw-8, gtpl449, gtpl453, atl-gw-22, a2b adenosine receptor antagonists, adenosine/ortho-mcneil
N-(4-cyanophenyl)-2-[4-(2,6-dioxo-1,3-dipropyl-7H-purin-8-yl)phenoxy]acetamide
C26H26N6O4
AJBBEYXFRYFVNM-UHFFFAOYSA-N
6603931
CCCN1C2=C(C(=O)N(C1=O)CCC)NC(=N2)C3=CC=C(C=C3)OCC(=O)NC4=CC=C(C=C4)C#N

Specifications

MRS 1754
264622-58-4
n-4-cyanophenyl-2-4-2,6-dioxo-1,3-dipropyl-7h-purin-8-yl phenoxy acetamide, n-4-cyanophenyl-2-4-2,3,6,7-tetrahydro-2,6-dioxo-1,3-dipropyl-1h-purin-8-yl phenoxy-acetamide, lopac-m-6316, d01qyq, d0ne5p, atl-gw-8, gtpl449, gtpl453, atl-gw-22, a2b adenosine receptor antagonists, adenosine/ortho-mcneil
CCCN1C2=C(C(=O)N(C1=O)CCC)NC(=N2)C3=CC=C(C=C3)OCC(=O)NC4=CC=C(C=C4)C#N
486.532
486.52
50mg
C26H26N6O4
AJBBEYXFRYFVNM-UHFFFAOYSA-N
N-(4-cyanophenyl)-2-[4-(2,6-dioxo-1,3-dipropyl-7H-purin-8-yl)phenoxy]acetamide
6603931
>98%
Videos