Learn More
MPPG, Tocris Bioscience™
Group III and group II mGlu antagonist; more selective for group III than group II
Brand:  Tocris (Bio-Techne) 085350
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13448073
  • £520.38 / 50mg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
Potent antagonist of L-AP4-induced effects in rat spinal cord, thalamic and hippocampal neurons, showing selectivity over (1S,3S)-ACPD-induced effects. Potent antagonist of mGlu receptors linked to adenylyl cyclase in adult rat cortical slices.
Specifications
Chemical Identifiers
169209-65-8
245.171
mppg, 2-amino-2-4-phosphonophenyl propanoic acid, chembl86508, d0d2gl, mppg nmr , solid, rs-?-methyl-4-phosphonophenylglycine, rs-, a-methyl-4-phosphonophenylglycine, 2-amino-2-4-phosphono-phenyl-propionic acid, 2-amino-2-4-phosphono-phenyl-propionic acid rs-mppg
2-amino-2-(4-phosphonophenyl)propanoic acid
C9H12NO5P
PAONCRJPUQXPRW-UHFFFAOYSA-N
3972752
CC(C1=CC=C(C=C1)P(=O)(O)O)(C(=O)O)N
Specifications
MPPG
C9H12NO5P
PAONCRJPUQXPRW-UHFFFAOYSA-N
2-amino-2-(4-phosphonophenyl)propanoic acid
3972752
>98%
169209-65-8
mppg, 2-amino-2-4-phosphonophenyl propanoic acid, chembl86508, d0d2gl, mppg nmr , solid, rs-?-methyl-4-phosphonophenylglycine, rs-, a-methyl-4-phosphonophenylglycine, 2-amino-2-4-phosphono-phenyl-propionic acid, 2-amino-2-4-phosphono-phenyl-propionic acid rs-mppg
CC(C1=CC=C(C=C1)P(=O)(O)O)(C(=O)O)N
245.171
245.17