missing translation for 'onlineSavingsMsg'
Learn More

MPP dihydrochloride, Tocris Bioscience™

Highly selective ERα antagonist

Brand:  Tocris (Bio-Techne) 199150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13440364

  • £686.72 / 50mg

Estimated Shipment
09-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective, high affinity silent antagonist at ERα receptors. Displays > 200-fold selectivity for ERα over ERβ. Ki values are 2.7 and 1800 nM at ERα and ERβ receptors respectively.
Specifications

Chemical Identifiers

911295-24-4
542.501
mpp dihydrochloride, 1,3-bis-4-hydroxyphenyl-4-methyl-5-4-2-piperidinylethoxy phenol-1h-pyrazole dihydrochloride, mpp dihydrochloride hydrate, methyl-piperidino-pyrazole hydrate, 1,3-bis 4-hydroxyphenyl-4-methyl-5-4-2-piperidinylethoxy phenol-1h-pyrazole dihydrochloride, mpp dihydrochloride hydrate hplc , white powder, 1,3-bis 4-hydroxyphenyl-4-methyl-5-4-2-piperidinylethoxy phenol-1h-pyrazole dihydrochloride hydrate, 4-1-4-hydroxyphenyl-4-methyl-5-4-2-piperidin-1-yl ethoxy phenyl-1,2-dihydro-3h-pyrazol-3-ylidene cyclohexa-2,5-dien-1-one-hydrogen chloride 1/2, 4-3-4-hydroxyphenyl-4-methyl-5-4-2-piperidin-1-yl ethoxy phenyl pyrazol-1-yl phenol dihydrochloride
4-[2-(4-hydroxyphenyl)-4-methyl-3-[4-(2-piperidin-1-ylethoxy)phenyl]-1H-pyrazol-5-ylidene]cyclohexa-2,5-dien-1-one;dihydrochloride
C29H33Cl2N3O3
SMPRQALBFACGGO-UHFFFAOYSA-N
45073474
CC1=C(N(NC1=C2C=CC(=O)C=C2)C3=CC=C(C=C3)O)C4=CC=C(C=C4)OCCN5CCCCC5.Cl.Cl

Specifications

MPP dihydrochloride
911295-24-4
mpp dihydrochloride, 1,3-bis-4-hydroxyphenyl-4-methyl-5-4-2-piperidinylethoxy phenol-1h-pyrazole dihydrochloride, mpp dihydrochloride hydrate, methyl-piperidino-pyrazole hydrate, 1,3-bis 4-hydroxyphenyl-4-methyl-5-4-2-piperidinylethoxy phenol-1h-pyrazole dihydrochloride, mpp dihydrochloride hydrate hplc , white powder, 1,3-bis 4-hydroxyphenyl-4-methyl-5-4-2-piperidinylethoxy phenol-1h-pyrazole dihydrochloride hydrate, 4-1-4-hydroxyphenyl-4-methyl-5-4-2-piperidin-1-yl ethoxy phenyl-1,2-dihydro-3h-pyrazol-3-ylidene cyclohexa-2,5-dien-1-one-hydrogen chloride 1/2, 4-3-4-hydroxyphenyl-4-methyl-5-4-2-piperidin-1-yl ethoxy phenyl pyrazol-1-yl phenol dihydrochloride
CC1=C(N(NC1=C2C=CC(=O)C=C2)C3=CC=C(C=C3)O)C4=CC=C(C=C4)OCCN5CCCCC5.Cl.Cl
542.501
542.5
50mg
C29H33Cl2N3O3
SMPRQALBFACGGO-UHFFFAOYSA-N
4-[2-(4-hydroxyphenyl)-4-methyl-3-[4-(2-piperidin-1-ylethoxy)phenyl]-1H-pyrazol-5-ylidene]cyclohexa-2,5-dien-1-one;dihydrochloride
45073474
>98%
Videos
Documents