Learn More
Tocris Bioscience™ MPEP hydrochloride
Potent mGlu5 antagonist; also positive allosteric modulator of mGlu4 receptors
Brand:  Tocris Bioscience™ 1212/10
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13438893
  • £145.53 / 10mg
Estimated Shipment date
from Supplier 16-12-2019
Description
Description
Potent and highly selective non-competitive antagonist at the mGlu5 receptor subtype (IC50 = 36 nM) and a positive allosteric modulator at mGlu4 receptors. Centrally active following systemic administration in vivo. Reverses mechanical hyperalgesia in the inflamed rat hind paw. Also available as part of the Group I mGlu Receptor Tocriset™ .
Specifications
Specifications
2-Methyl-6-(phenylethynyl)pyridine hydrochloride
>99%
Observed MW: 229.71
Soluble to 100mM in DMSO
219911-35-0
C14H11N.HCl