missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Yeast Hydrolysate Enzymatic

Autolyzed Brewers yeast

£61.27 - £986.86

Specifications

Content And Storage 15-30°C
Form Tan powder
Source Brewers Yeast
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11437541
View Documents Promotion Details
MP Biomedicals™
0210330405
5 lb.
£221.54
5lb
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
11447541
View Documents Promotion Details
MP Biomedicals™
0210330425
25 lb.
£986.86
25lb
View alternative products
Save Up to 
»
Estimated Shipment: 09-08-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
11427541
View Documents Promotion Details
MP Biomedicals™
0210330401
1 lb.
£61.27
1lb
View alternative products
Save Up to 
»
Estimated Shipment: 09-08-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Yeast Hydrolysate Enzymatic is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast.

Specifications

Specifications

15-30°C
Tan powder
Brewers Yeast
Videos
SDS
Documents

Documents

Product Certifications