MP Biomedicals RPM™ Kit


Overview
Brand: MP Biomedicals

Manufacturer Part Number: 112070500

UNSPSC: 41116104

Code: 50

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 11383149

Quantity Price
1 £ 619.3 / 300 preps
Estimated Shipment
15-05-2017
Add to basket


Description and Specification

Specification

Quantity 300 Preparations
Includes Pre-Lysis, Alkaline Lysis, Neutralizing, Glassmilk spin buffers, wash soln. concentrate
For Use With (Application) For rapid isolation, purification of double-stranded plasmid DNA from bacterial culture
Product Type RPM Kit