MP Biomedicals RPM™ Kit


Overview
Brand: MP Biomedicals

Manufacturer Part Number: 112070400

RPM® KIT 120 preps

UNSPSC: 41116104

Code: 50

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 11373149

Quantity Price
1 £ 255.2 / 120 preps
Estimated Shipment
04-04-2017
Add to basket


Description and Specification

Specification

For Use With (Application) For rapid isolation, purification of double-stranded plasmid DNA from bacterial culture
Size 120 Preparations
Includes Pre-Lysis, Alkaline Lysis, Neutralizing, Glassmilk spin buffers, wash soln. concentrate
Product Type RPM Kit
Quantity 120 Preparations
Product Name RPM Kit