missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ RapidPURE™ Plasmid Mini Kit

For use in routine molecular biology applications such as fluorescent and radioactive sequencing and cloning

Brand:  MP Biomedicals™ 112066200

Code : 50

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11323149

  • POA / 60 preps

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Description

Description

  • The Kit is designed to isolate up to 20μg of plasmid DNA from 1.5mL of an overnight bacterial culture.
  • All steps can be carried out in either a microcentrifuge or vacuum manifold.
Specifications

Specifications

Isolate up to 20ug of plasmid DNA from 1.5mL of an overnight bacterial culture
Plasmid isolation kit
60 Preps
Store at Room temperature (15° to 30°C)
Videos