missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Proteinase K
Suitable for both protein and nucleic acid isolation
£53.78 - £1169.18
Specifications
For Use With (Application) | Protein Structure Analysis |
---|---|
Molecular Weight (g/mol) | ∽30 |
Name | Proteinase K |
pH Range | 7.5 to 12 |
Purification Method | Chromatography |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484822
|
MP Biomedicals™
0219350491 |
1mg |
£1169.18
1g |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11444822
|
MP Biomedicals™
0219350425 |
25mg |
£68.79
25mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11454822
|
MP Biomedicals™
0219350480 |
100mg |
£53.78
100mg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11464822
|
MP Biomedicals™
0219350483 |
250mg |
£323.30
250mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11474822
|
MP Biomedicals™
0219350490 |
500mg |
£560.74
500mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Proteinase K, [39450-01-6]
Proteolytic Activity: >30mAnson Units/mg
Unit Definition: One Anson unit is the amount of enzyme which liberates 1 μmol of Folin-positive amino acids per minute at pH 7.5 and 35°C, using hemoglobin as substrate
- From Tritirachium album
- Chromatographically purified
- Exhibits proteolytic activity on proteins, peptides, glycoproteins, amides and esters.
- Active with nitroanilides of amino acids with protected amino groups, excluding arginine.
- Useful in the isolation of DNA and RNA, in the analysis of membrane structures and protein structure.
Specifications
Protein Structure Analysis | |
Proteinase K | |
Chromatography | |
4°C |
∽30 | |
7.5 to 12 | |
Tritirachium album Limber | |
Lyophilized |