missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Pepstatin A
Synthetic inhibitor for pepsin, renin, cathepsin D, and other acid proteases.
£66.67 - £1855.00
Specifications
For Use With (Application) | Enzyme Assay, Inhibition Assay, Molecular Biology, Protein Purification, Sample Preparation |
---|---|
Molecular Weight (g/mol) | 685.91 |
Name | Pepstatin A |
Storage Requirements | 0°C |
Form | Powder |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11485322
|
MP Biomedicals™
0219536805 |
5mg |
£66.67
5mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11405332
|
MP Biomedicals™
0219536825 |
25mg |
£225.78
25mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11495322
|
MP Biomedicals™
0219536810 |
10mg |
£177.02
10mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11425332
|
MP Biomedicals™
0219536883 |
250mg |
£1855.00
250mg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Pepstatin A, [26305-03-3]; Isoval-Val-Val-Sta-Ala-Sta
C34H63N5O9, F.W. 685.9; Melting Point: 235-237°C
Specifications
Enzyme Assay, Inhibition Assay, Molecular Biology, Protein Purification, Sample Preparation | |
Pepstatin A | |
Powder |
685.91 | |
0°C |