missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysing Matrix Y

For lysis of gram positive and negative bacteria, fungal tissue and spores, yeasts, molds, filamentous algaes and biofilms

Brand:  MP Biomedicals™ 116960500

Additional Details : Weight : 0.45300kg

 View more versions of this product

Product Code. 15282840

  • £1207.00 / Pack of 500


Estimated Shipment
04-05-2018
Add to basket

Description

Description

  • Impact-resistant 2mL tube contains 0.5mm diameter Yttria-Stabilized Zirconium Oxide beads
  • Specific garvity: 6.02

Applications:

Nucleic Acid Isolation and Purification

Specifications

Specifications

Dnase-free
Lysing matrix Y
RNase-free
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis
500 x 2mL Tubes
Videos