missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ LB-Agar, Capsules, Molecular Biology Certified™
Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium.
£0.05 - £234.26
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11323259
|
MP Biomedicals™
113002211 |
227 g |
£89.78
227g |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
11343259
|
MP Biomedicals™
113002221 |
454 g |
£234.26
454g |
Estimated Shipment: 16-04-2024
Log in to see stock. |
|||||
15712612
|
MP Biomedicals™
113002201 |
11 g |
£0.05
11g |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
- Nutritionally rich medium originally developed for growth and maintenance of E. coli strains
- Eliminates the waste, inaccuracies, and mess associated with weighing out bulk powder
- Contains essential electrolytes for transport and osmotic balance, due to NaCl component
Specifications
15°C to 30°C | |
Capsule |