Learn More
MP Biomedicals™ Geneclean™ Turbo Kit, 100 Preps
For nucleic acid isolation and purification
Brand: MP Biomedicals™ 111102400
Code : 50
Additional Details : Weight : 0.30000kg Transport : UN number : 2922 Chem class : 8 Pack group : III
Description
GeneClean™ Turbo Kit is a flexible kit designed for purification of DNA fragments of sizes 0.1 kb to 300 kb from TAE or TBE buffered agarose gels, PCR reactions, and other enzymatic solutions. It purifies larger fragments due to less manipulation of bound DNA. Process with either vacuum or centrifugation & save time with patented Turbo filter design.
Application Notes
GENECLEAN™ Turbo Kit is used for purification of DNA fragments of sizes 0.1 kb to 300 kb from agarose gels, PCR reactions, and other enzymatic solutions. Purified DNA can be used in enzymatic reactions, conventional or automated sequencing, cloning, and microarray analysis.
Usage Statement
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.
Key Applications
Purification of DNA fragments from agarose gels; PCR reactions | Elimination of proteins from enzymatic reactions
Specifications
Geneclean Imbedded Turbo Spin Column | |
Nucleic Acid isolation and purification | |
110mL Geneclean™ Turbo™ Salt solution, 11mL Geneclean™ Turbo™ wash concentrate, 100 Geneclean™ Turbo™ cartridges, 100 Geneclean™ | |
100 Preps |
Safety and Handling
- geneclean turbo kit
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : guanidine thiocyanate