missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Geneclean™ Spin Kit 200 bp to 300 kb
Used to purify fragments of DNA from 200 bp to 300 kbp from solutions and agarose gels. The GENECLEAN™ SPIN kit uses SPIN filters, which prevent silica particle carry-over into cleaned DNA. Glassmilk™ slurry allows flexibility of reaction size and volume.
£203.52 - £859.66
Specifications
CAS | 593-84-0 |
---|---|
Format | Kit |
Sample Type | Enzymatic Reactions, Solutions, Agarose Gels |
For Use With (Application) | Isolate DNA from TAE or TBE buffered agarose gels and solutions |
Includes | SPIN filters |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11313119
|
MP Biomedicals™
111101200 |
50 Preps |
£203.52
50 preps |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11333119
|
MP Biomedicals™
111101400 |
100 Preps |
£334.96
100 preps |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11363119
|
MP Biomedicals™
111101600 |
300 Preps |
£859.66
300 preps |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- The GENECLEAN™ SPIN kit simplifies the process of purifying DNA into three easy steps: Bind, Wash and Elute
- Ethanol precipitation is never required and purified DNA is immediately ready for a wide variety of downstream applications
- Examples of applications are desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearized from uncut vector and isolation of PCR products away from genomic DNA
Specifications
593-84-0 | |
Enzymatic Reactions, Solutions, Agarose Gels | |
SPIN filters | |
Spin Kit | |
DNA Clean-Up Kit |
Kit | |
Isolate DNA from TAE or TBE buffered agarose gels and solutions | |
Geneclean SPIN New Wash pH Range: 7.4 to 7.7, GC Spin Elution Solution pH Range: 6.8 to 7.2 | |
Up to ~300 mg Gel Slice or up to ~300 μL DNA Solution | |
Gels and Solutions |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.