missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Collagenase, Type 4

From Clostridium histolyticum

£87.03 - £293.62

Specifications

Concentration >160 U/mg
Color Brown
For Use With (Application) For tissue dissociation combined with other Enzymes such as Elastase, Trypsin/Papain
Content And Storage 4°C
Source Clostridium histolyticum
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11410982
View Documents Promotion Details
MP Biomedicals™
0219511080
100 mg
£87.03
100mg
View alternative products
Save Up to 
»
Estimated Shipment: 08-07-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
11420982
View Documents Promotion Details
MP Biomedicals™
0219511090
500 mg
£293.62
500mg
View alternative products
Save Up to 
»
Estimated Shipment: 19-07-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pancreatic Islet Isolation Grade. Suggested for pancreatic islet isolation. Prepared to contain lower tryptic activity levels. Activity: ≥160U per mg solid. Unit Definition: One unit liberates peptides from collagen equivalent in ninhydrin color to 1.0μmol of L-leucine in 5 hr. at pH 7.4 and 37°C in presence of calcium ions. Dialyzed, lyophilized.

Specifications

Specifications

>160 U/mg
For tissue dissociation combined with other Enzymes such as Elastase, Trypsin/Papain
Clostridium histolyticum
Cysteine, EDTA or o-phenanthroline, alpha2-macroglobulin - a large plasma glycoprotein, alpha2-macroglobulin, alpha1-antitrypsin
Brown
4°C
Collagenase Type 4
Lyophilized Powder
Videos
SDS
Documents

Documents

Product Certifications