missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CIRCLEGROW™, Powder
Superior plasmid yield through enhanced cell growth
£75.26 - £937.04
Specifications
Content And Storage | 15° to 30°C |
---|---|
Product Type | Bacterial Growth Medium |
Sterility | Sterile |
pH | 6.9 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11323189
|
MP Biomedicals™
113000122 |
454 g |
£125.08
454g |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11333199
|
MP Biomedicals™
113000165 |
10 x 0.5 L |
£84.16
5 L |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11353199
|
MP Biomedicals™
113000175 |
10 x 1 L |
£132.50
1 L |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11303199
|
MP Biomedicals™
113000152 |
5 kg |
£937.04
5kg |
N/A | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
11393179
|
MP Biomedicals™
113000112 |
227 g |
£75.26
227g |
Estimated Shipment: 18-04-2024
Log in to see stock. |
||||
11353189
|
MP Biomedicals™
113000132 |
908 g |
£232.14
908g |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11383189
|
MP Biomedicals™
113000142 |
2.3 kg |
£504.56
2.30kg |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Description
Perfect for overnight or weekend cultures, CIRCLEGROW Medium is a super-rich bacterial growth medium that produces up to three times more plasmid DNA than traditional growth medium. CIRCLEGROW Medium consists of a protein and amino acid source, biological buffer, and a complex carbon source that gets metabolized at a much slower rate than glucose, thus prolonging the cell cycle and preventing cell death during the log phase of growth.
- Cells grown with CIRCLEGROW Medium remain healthy for more than two days and continue to divide slowly after the log phase of the growth has been completed
- Reduce hands-on work and preparation time
- Cultures do not need to be constantly monitored by O.D. readings due to the incremental post-log cell growth
- Obtain high yields of plasmid DNA from smaller culture volumes, and confidently leave cultures overnight or over the weekend
- Capsules are the most convenient form of this unique growth media
- Pre-mixed CIRCLEGROW is also available in single-use pouches, capsules and sterile liquid
- MP113000165: Includes ten 20 g pouches. Add contents of one pouch to 500mL of purified water.
- MP113000175: Includes ten 40g pouches. Add contents of one pouch to 1L of purified water.
- MP113000185: Includes one hundred 20g pouches. Add contents of one pouch to 500mL of purified water.
Specifications
15° to 30°C | |
Bacterial Growth Medium | |
Sterile | |
6.9 |