missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Albumin Human 10%, Prepared from Fraction V human albumin

Supplied at a pH of 7.1 ±0.3 and contains 0.05% sodium azide as a preservative

Brand:  MP Biomedicals™ 08810171

Code : NEW

Additional Details : CAS Number : 70024-90-7 Weight : 0.05000kg

Product Code. 11474216

  • £310.58 / 50mL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • CAS: 70024-90-7
  • Purity: >96.0 %

Specifications

Specifications

65
7.1
Human Albumin
Liquid
Human Albumin Protein
50mL
0° to 5°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers