missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSLP Mouse Uncoated ELISA Kit

Uncoated ELISA Kit

Brand:  Invitrogen™ 88-7490-88

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15571157

  • £301.00 / Pack of 10


Estimated Shipment date
from Supplier 02-04-2020
Add to basket

Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer. 10X PBS ELISA Coating Buffer
Assay Diluent. 5X Concentrated
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for the dilution of antibodies and standards

Description

Description

TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.
Specifications

Specifications

Q9JIE6
Biotin
53603
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, Coating Buffer: 10X PBS ELISA Coating Buffer, Assay Diluent: 5X Concentrated, Detection Enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Solution, Certificate of Analysis: Lot-specific instructions for the dilution of antibodies and standards, 96-Well Plates: Corning Costar flat-bottom plates (included with Cat. Nos. ending in -22 and -86)
RUO
Serum, 100 μL|Supernatant, 100 μL
1 hr. 45 min.
Mouse
Thymic Stromal-Derived Lymphopoietin
10 x 96 Tests
Supernatant
16-2,000 pg/mL
ELISA
Colorimetric Microplate Reader
ELISA Kit
16 pg/mL
2°C to 8°C
24 hr. 30 min.
G0S19-1,LD78ALPHA,MIP-1-alpha,MIP1A,SCYA3
HRP
Mouse
Videos
Documents

For Research Use Only