missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse TGF beta 1
Recombinant Protein
Brand: Affymetrix eBioscience 14-8342-62
Code : NEW
Additional Details : Weight : 0.09500kg
Description
TGF beta-1 (TGFB1, Transforming Growth Factor Beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its pro-domain. Activation of TGF beta-1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF beta-1, an amino-terminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP. TGF beta-1 helps regulates proliferation, differentiation, adhesion, migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. TGF beta-1 is cleaved into a latency-associated peptide and a mature TGF beta-1 peptide, and is found in either a latent form composed of a TGFB1 homodimer, a LAP homodimer, and a latent TGFB1-binding protein, or in an active form composed of a TGF beta-1 homodimer. The mature peptide may also form heterodimers with other TGF beta family members. The gene for TGF beta-1 is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease and cystic fibrosis. Description: Mouse TGF beta 1 (Transforming Growth Factor beta 1) is a member of a superfamily of homologous, disulfide-linked, homodimeric proteins that regulate the proliferation and differentiation of normal and transformed cells. Human TGF beta 1 is a 25 kDa protein with each subunit containing 112 aa residues. TGF beta 1 is secreted from the many cell types in a complex with LAP (Latency Associated Peptide) and also with LTBP (latent TGF beta binding protein). Disassociation is thought to happen in the extracellular matrix via proteases and/or integrins. Regulatory T cells are a recently identified source of TGF beta 1 thought to be a key player in their suppressive functions. TGF beta 1 binds through TGF beta receptors (of which there is a large family). The protein is highly conserved across all mammalian species. Applications Reported: Recombinant mouse TGF-β1 is biologically active and can inhibit IL-4 induced proliferation of mouse HT-2 cells. Applications Tested: The ED50 of this protein, as measured by inhibition of IL-4 induced proliferation in HT-2 cells, is less than or equal to 40 pg/mL. This corresponds to a specific activity of greater than or equal to 2.5 x 10e7 Units/mg. Bioactivity: The ED50 of this protein, as measured by inhibition of IL-4 induced proliferation in HT-2 cells, is less than or equal to 40 pg/mL. This corresponds to a specific activity of greater than or equal to 2.5 x 10e7 units/mg. Endotoxin: Less than 0.01ng/ug cytokine as determined by the LAL assay. Purity: >95% as determined by SDS-PAGE. Molecular Weight: Mature TGFb1 polypeptide has a predicted molecular mass of 12,794. On reducing SDS-PAGE the protein migrates as a 13 kDa polypeptide and on non-reducing SDS page the cystine-linked homodimer migrates as a 25 kDa protein. Storage and handling: For best recovery, quick-spin vial prior to opening. Use in a sterile environment. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2um post-manufacturing filtered.Specifications
P04202 | |
Control, Functional Assay, Neutralization | |
21803 | |
Mouse TGF beta 1 | |
SDS-PAGE | |
-80°C | |
Less than 0.01ng/μg cytokine as determined by the LAL assay. | |
Tgfb1 | |
Recombinant Protein | |
Unconjugated | |
Mouse | |
Liquid | |
Affinity chromatography |
0.1mg/mL | |
protein with 0.1M glycine, 1% BSA and no preservative; pH 3.0 | |
Mature TGFb1 polypeptide has a predicted molecular mass of 12,794. On reducing SDS-PAGE the protein migrates as a 13kDa polypeptide and on non-reducing SDS page the cystine-linked homodimer migrates as a 25kDa protein. | |
3.0 | |
5μg | |
RUO | |
Mouse TGF beta 1 | |
The ED50 of this protein, as measured by inhibition of IL-4 induced proliferation in HT-2 cells, is less than or equal to 40pg/mL. This corresponds to a specific activity of greater than or equal to 2.5 x 107 units/mg. | |
Mouse | |
Ms | |
Recombinant | |
Recombinant |