Thermo Scientific™ Pierce™ Mouse Soluble ICAM-1 (CD54) ELISA Kits

Measure levels of mouse soluble ICAM-1 (CD54) in mouse serum, plasma (EDTA) and cell culture supernatant samples using 96-well plates.

Overview
Brand: Thermo Scientific™ Pierce™

Manufacturer Part Number: EMICAM12

UNSPSC: 41116126

Code: PN

Additional Details:
Additional Details: Weight: 0.10000kg



Product Code. 10638084

Quantity Price
1 £ 443.0 / 1 set
Estimated Shipment date
from Supplier 07-04-2017
Add to basket


Description and Specification

Specification

Precision Intra-assay CV <10%; inter assay CV <10%
Sensitivity <5pg/mL
Measurement Absorbance at 450nm minus absorbance at 550nm
Formulation Complete ELISA kit with coated 96-well plate
Kit Contents 30X Wash Buffer, 2 x 50mL
Assay Range 25.6 to 1000pg/mL
Sufficient For 96 microplate wells
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Specificity No significant cross reaction with rat or human sICAM 1
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Target Mouse soluble ICAM-1 (intercellular adhesion molecule 1, also called CD54)
Description Mouse Soluble ICAM-1 (CD54) ELISA Kit
Sample Size 50µL of 100-fold diluted samples per well
Total Assay Time 3.5 hr.
Quantity 2 plate kit

These mouse sICAM-1 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of soluble ICAM-1 (intercellular adhesion molecule 1, also called CD54) in samples at 50μL per well. Also included are calibrated recombinant protein standard, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Mouse soluble ICAM-1 (intercellular adhesion molecule 1, also called CD54)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 25.6 to 1000ng/mL

  • Sensitivity: < 5ng/mL

  • Precision: Intra-assay CV < 10%; inter assay CV < 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3.5 hours

  • Sample Size: 50μL of 100 fold diluted samples per well

  • Specificity: No significant cross reaction with rat or human sICAM 1

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Documentation

Technical documents